Learn More
CD56 Mouse anti-Human , BB700, Clone: NCAM16.2 (also known as NCAM 16), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD56 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | NCAM16.2 (also known as NCAM 16) |
| Conjugate | Brilliant Blue 700 (BB700) |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 |
| Host Species | Mouse |
| Immunogen | Immunoaffinity-enriched adult human brain NCAM |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.