Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD56 Mouse anti-Human, Porcine, R-PE, Clone: MEM 188, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 945609
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD56 | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Acute myelogenous leukemia cell line KG-1 | |
RUO | |
Human, Pig | |
IgG2a |
Flow Cytometry | |
MEM 188 | |
R-PE | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction