Learn More
CD56 Mouse anti-Human, Porcine, SPRD (R-phycoerythrin/Cyanine 5), Clone: MEM 188, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
Specifications
| Antigen | CD56 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM 188 |
| Concentration | Lot specific |
| Conjugate | SPRD |
| Formulation | Supplied in PBS/NaN3 and a stabilizing agent |
| Host Species | Mouse |
| Immunogen | Acute myelogenous leukemia cell line KG-1 |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.