Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD56/NCAM-1 Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1149P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1149P 1 mL Concentrated 200μg/mL
MS1149PABX 200 μL Concentrated 1mg/mL
MS1149R7 7 mL Prediluted -
MS1149P0 100 μL Concentrated 200μg/mL
MS1149P1ABX 100 μL Concentrated 1mg/mL
MS1149P1 500 μL Concentrated 200μg/mL
MS1149R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1149P Supplier Epredia Supplier No. MS1149P
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD56/NCAM-1 Ab-4, Mouse Monoclonal Antibody.

Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule which is glycosylated or sialylated to produce the mature species. NCAM (CD56) is reported to express on most neuroectodermal derived cell lines, tissues, and neoplasms such as retinoblastoma, medullblastoma, astrocytoma, and neuroblastoma. It is also expressed on some mesodermally derived tumors such as rhabdomyosarcoma and also on natural killer cells.

Host Species: Mouse

Clone: 56C04; same as 123A8

Isotype: IgG1/κ

Species Reactivity: Human. Others not known

Epitope: Not determined

Immunogen: Membrane preparation of a small cell lung carcinoma

Molecular Weight: 180, 145 and 125 kDa

Positive Control: Neuroblastoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD56 Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 56C04; same as 123A8
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Membrane preparation of a small cell lung carcinoma
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.