Learn More
Description
Host Species: Mouse
Clone: 56C04; same as 123A8
Isotype: IgG1/κ
Species Reactivity: Human. Others not known
Epitope: Not determined
Immunogen: Membrane preparation of a small cell lung carcinoma
Molecular Weight: 180, 145 and 125 kDa
Positive Control: Neuroblastoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD56 Ab-4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 56C04; same as 123A8 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Membrane preparation of a small cell lung carcinoma |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.