Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD56 (NCAM-1), Mouse Anti-Human, RB613, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
$148.00 - $370.00
Specifications
Antigen | CD56 (NCAM-1) |
---|---|
Clone | B159 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB613 |
Description
CD56 (NCAM-1) Monoclonal antibody specifically detects CD56 (NCAM-1) in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow CytometrySpecifications
CD56 (NCAM-1) | |
Flow Cytometry | |
RB613 | |
Mouse | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 | |
Human NK Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
B159 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
4684 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title