Learn More
BD Horizon™ CD56 (NCAM-1) Mouse, anti-Human, RY586, Clone: B159, BD Horizon™
Shop All BD Biosciences ProductsDescription
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Specifications
Specifications
| Antigen | CD56 (NCAM-1) |
| Applications | Flow Cytometry |
| Clone | B159 |
| Conjugate | RY586 |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.