Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD57 Antibody (NK-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | beta-1,3-Glucuronyltransferase 1/B3GAT1 |
---|---|
Clone | NK-1 |
Concentration | 1.0 mg/mL |
Dilution | Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 - 1 ug/ml |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247789A
![]() |
Novus Biologicals
NBP2477890.1MG |
0.1 mg |
Each for $646.00
|
|
|||||
NBP247789B
![]() |
Novus Biologicals
NBP2477890.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
CD57 Monoclonal specifically detects CD57 in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
beta-1,3-Glucuronyltransferase 1/B3GAT1 | |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Immunology | |
PBS with No Preservative | |
27087 | |
Human peripheral blood mononuclear cells | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
NK-1 | |
Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 - 1 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat (Negative) | |
Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase | |
B3GAT1 | |
IgM κ | |
Protein A or G purified | |
Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title