Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD57 Antibody (NK1/7565), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32027520UG
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CD57 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
Recombinant fragment (around aa1-200) of human CD57 protein (exact sequence is proprietary) | |
20 μg | |
Cancer, Cell Biology, Immunology | |
27087 | |
Store at 4&drg;C. Do not freeze. | |
IgG2 λ |
Immunohistochemistry (Paraffin) | |
NK1/7565 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction