Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD57 Monoclonal Antibody (G539.1), QED Bioscience™
SDP

Catalog No. 502277997
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7997 Supplier Leinco Technologies Inc Supplier No. 34054100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CD57
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G539.1
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation tris with 1% BSA and 0.1% sodium azide; pH 7.3-7.7
Gene B3GAT1
Gene Accession No. Q9P2W7
Gene Alias HNK-1; Leu-7
Host Species Mouse
Immunogen Recombinant human CD57.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.