Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD57 Monoclonal Antibody (NK1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA512008
Description
MA5-12008 targets CD57 in FACS and IHC (P) applications and shows reactivity with Human samples. The MA5-12008 immunogen is human peripheral blood mononuclear cells.
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).
Specifications
CD57 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q9P2W7 | |
B3GAT1 | |
Human peripheral blood mononuclear cells | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgM κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
NK1 | |
Unconjugated | |
B3GAT1 | |
HNK-1; Leu-7 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
27087 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction