Learn More
CD57 (Natural Killer Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: NK1
Isotype: IgM/κ
Species Reactivity: Human. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Human peripheral blood mononuclear cells
Molecular Weight: 110 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
Specifications
| Antigen | CD57 (Natural Killer Cell Marker) Ab-1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | NK1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human peripheral blood mononuclear cells |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.