Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD57 (Natural Killer Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS136P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS136P1 500 μL Concentrated 200μg/mL
MS136P1ABX 100 μL Concentrated 1mg/mL
MS136PABX 200 μL Concentrated 1mg/mL
MS136R7 7 mL Prediluted -
MS136P 1 mL Concentrated 200μg/mL
MS136R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS136P1 Supplier Epredia Supplier No. MS136P1
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments.

CD57 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.

Host Species: Mouse

Clone: NK1

Isotype: IgM/κ

Species Reactivity: Human. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Human peripheral blood mononuclear cells

Molecular Weight: 110 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Specifications

Antigen CD57 (Natural Killer Cell Marker) Ab-1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NK1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.