Learn More
Epredia™ Lab Vision™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: HNK-1; same as Leu-7
Isotype: IgM/κ
Species Reactivity: Human. Others not known.
Immunogen: Membrane extracts of the HSB-2 T-lymphoblastoid cell line
Molecular Weight: 110 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD57 (Natural Killer Cell Marker) Ab-2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HNK-1; same as Leu-7 |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | Membrane extracts of the HSB-2 T lymphoblastoid cell line |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.