Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B3GAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597275 Supplier Invitrogen™ Supplier No. PA597275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHKDEGSDPR RETPPGADPR EYCTSDRDIV EVVRTEYVYT RPPPWSDTLP TIHVVTPTYS RPVQKAELTR MANTLLHVPN LHWLVVEDAP RRTPLTARLL RDTGLNYTHL HVETPRNYKL RGDARDPRIP RGTMQRNLAL RWLRETFPRN SSQPGVVYFA DDDNTYSLEL FEEMRSTRRV SVWPVAFVGG LRYEAPRVNG AGKVVGWKTV FDPHRPFAID MAGFAVNLRL ILQRSQAYFK LRGVKGGYQE SSLLRELVTL NDLEPKAANC TKILVWHTRT EKPVLVNEGK KGFTDPSVEI; Positive Samples: SW480, HT-1080, BT-474; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B3GAT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene B3GAT1
Gene Accession No. Q9P2W7
Gene Alias 0710007K08Rik; AI846286; B3GAT1; Beta-1,3-glucuronyltransferase 1; beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P); CD57; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; GLCATP; glcAT-P; GLCUATP; GlcUAT-P; Glucuronosyltransferase P; glucuronosyltransferase-P; Hnk1; Hnk-1; LEU7; LEU7 antigen; NK1; NK-1; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Symbols B3GAT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 35-334 of human B3GAT1 (NP_4733661).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.