Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD57 Recombinant Mouse Monoclonal Antibody (PD00-21)
GREENER_CHOICE

Catalog No. FEMA550650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550650 Supplier Invitrogen™ Supplier No. MA550650
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Positive control: Human cervical cancer, human Hodgkin's lymphoma tissue, human tonsil tissue. Predicted band size: 38 kDa Subcellular Location: Golgi apparatus membrane, Secreted, Endoplasmic reticulum membrane.

CD57, also known as HNK-1 and Leu-7, is an oligosaccharide antigenic determinant present on a variety of polypeptides, lipids, and chondroitin sulphate proteoglycans. Its function is poorly understood. CD57 is expressed on a subset of natural killer (NK) cells and NK T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD57
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Multiplex Immunohistochemistry
Classification Recombinant Monoclonal
Clone PD00-21
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene B3GAT1
Gene Accession No. Q9P2W7
Gene Alias HNK-1; Leu-7
Gene Symbols B3GAT1
Host Species Mouse
Immunogen Synthetic peptide.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.