Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD58/LFA-3 Antibody (085), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290096MFV450
Description
CD58/LFA-3 Monoclonal antibody specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
CD58/LFA-3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
085 | |
Western Blot, Immunoprecipitation | |
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD58/LFA-3 (Accession#: AAH05930.1; Met1-Arg215). | |
0.1 mL | |
Cell Biology, Cellular Markers, Immunology | |
965 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction