Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD58/LFA-3 Antibody (MEM-63), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1201420C
This item is not returnable.
View return policy
Description
CD58/LFA-3 Monoclonal antibody specifically detects CD58/LFA-3 in Human, Porcine samples. It is validated for Flow CytometrySpecifications
CD58/LFA-3 | |
Monoclonal | |
DyLight 650 | |
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
NALM-6 human pre-B cell line | |
0.1 mL | |
Cell Biology, Cellular Markers, Immunology | |
965 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
MEM-63 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction