Learn More
CD58/LFA-3 Antibody (MM0176-8G24), Alexa Fluor™ 750, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212172AF750
Catalog No. NP212172A70
Only null left
Add to Cart
Description
Description
CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
CD58/LFA-3
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CD58
Human recombinant CD58/LFA-3 (extracellular domain)
Primary
Human
IgG2
Western Blot, Flow Cytometry
MM0176-8G24
Western Blot, Flow Cytometry
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3
Mouse
0.1 mL
965
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.