Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD58/LFA-3 Antibody (MM0176-8G24) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.72 - $554.59
Specifications
| Antigen | CD58/LFA-3 |
|---|---|
| Clone | MM0176-8G24 |
| Concentration | LYOPH |
| Applications | CyTOF, Western Blot, Flow Cytometry |
| Classification | Monoclonal |
Description
CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| CD58/LFA-3 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
| CD58 | |
| Protein G purified |
| MM0176-8G24 | |
| CyTOF, Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology | |
| P19256 | |
| 965 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title