Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD58/LFA-3 Antibody (MM0176-8G24), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212172UV
Description
CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD58/LFA-3 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD58 | |
Human recombinant CD58/LFA-3 (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
MM0176-8G24 | |
Western Blot, Flow Cytometry | |
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
Mouse | |
Protein G purified | |
Cell Biology, Cellular Markers, Immunology | |
965 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction