Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD58/LFA-3 Rabbit anti-Human, Clone: 83, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | CD58/LFA-3 |
---|---|
Clone | 83 |
Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
CD58/LFA-3 | |
Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
Recombinant Human CD58/LFA-3 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
83 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers, Immunology | |
0.2 um filtered solution in PBS | |
965 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title