Learn More
Invitrogen™ CD58 Monoclonal Antibody (MEM-63), Biotin

Description
This antibody reacts with CD58 (LFA-3), a 40-70 kDa extracellular membrane glycoprotein distributed over many tissues, leukocytes, erythrocytes, endothelial cells, epithelial cells and fibroblasts.
Specifications
Specifications
| Antigen | CD58 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-63 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | TBS with 15mM sodium azide; pH 8 |
| Gene | CD58 |
| Gene Accession No. | P19256 |
| Gene Alias | Ag3; CD305; CD58; CD58 antigen; CD58 antigen, (lymphocyte function-associated antigen 3); CD58 molecule; FLJ23181; FLJ43722; LAIR1; LAIR-1; LFA3; LFA-3; Lymphocyte function-associated antigen 3; membrane receptor; RP5-1086K13.2; surface glycoprotein LFA-3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.