Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD58 Mouse, anti-Human, R718, Clone: 1C3, BD OptiBuild™

Catalog No. BDB752136 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752136 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752136 Supplier BD Biosciences Supplier No. 752136
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1C3 (AICD58.6) monoclonal antibody specifically binds to CD58. CD58 is a a 60-70 kDa glycoprotein member of the immunoglobulin superfamily. CD58, also referred to as the lymphocyte function-associated antigen-3 (LFA-3), has a wide tissue distribution, being expressed on both hematopoietic and non-hematopoietic cells, including endothelial cells and fibroblasts. There are two isoforms of CD58: a glycosylphosphatidylinositol (GPI)-linked form and a transmembrane form. Both isoforms may be expressed on the same cell type. Erythrocytes, however, only express the GPI-linked isoform. CD58 interacts with CD2 during cell adhesion. This binding can enhance antigen-specific T-cell activation. This interaction can also play a role in cell-mediated cytotoxicity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD58
Applications Flow Cytometry
Classification Monoclonal
Clone 1C3
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.