Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD59 Antibody (2491C), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1987H
Description
CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CD59 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 966 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 2491C | |
| Flow Cytometry, CyTOF-ready | |
| 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein | |
| Mouse myeloma cell line, NS0-derived recombinant human CD59, Leu26-Asn102, Accession # P13987 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction