Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD59 Antibody (YTH53.1) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | CD59 |
---|---|
Clone | YTH53.1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:5-1:10, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017730
![]() |
Novus Biologicals
NB100655370.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065537
![]() |
Novus Biologicals
NB10065537 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CD59 Monoclonal specifically detects CD59 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CD59 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rat | |
Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers | |
P13987 | |
CD59 | |
IgG2b | |
Protein G purified | |
NB100-65537 recognizes the human CD59 cell surface antigen, an 18-20kD GPI linked glycoprotein broadly expressed by human leucocytes and erythrocytes. Clone YTH53.1 has been shown to block the normal function of CD59. Removal of sodium azide is recommended prior to use in functional assays. CD59 is expressed by erythrocytes, and therefore staining in whole blood systems requires a higher concentration of antibody. |
YTH53.1 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:5-1:10, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
966 | |
Human Peripheral Blood T cells | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title