Learn More
Invitrogen™ CD59 Recombinant Rabbit Monoclonal Antibody (5W9W7)

Description
Immunogen sequence: DACLITKAGL QVYNKCWKFE HCNFNDVTTR LRENELTYYC CKKDLCNFNE QLENGGTSLS EKTVLLLVTP FLAAAWSLHP.
Specifications
Specifications
| Antigen | CD59 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 5W9W7 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CD59 |
| Gene Accession No. | P13987 |
| Gene Alias | 16.3A5; 1F5; 1F5 antigen; 20 kDa homologous restriction factor; AA987121; Cb59b molecule; CD59; CD59 antigen; CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344); CD59 glycoprotein; CD59 molecule; CD59 molecule (CD59 blood group); CD59 molecule, complement regulatory protein; Cd59a; CD59a antigen; CD59A glycoprotein; Cd59b; CD59b antigen; CD59B glycoprotein; CD59b moleucle, complement regulatory protein; complement inhibitor CD59; complement regulatory protein; EJ16; EJ30; EL32; G344; HRF20; HRF-20; human leukocyte antigen MIC11; Ly-6-like protein; lymphocytic antigen CD59/MEM43; MACIF; MAC-inhibitory protein; MACIP; MAC-IP; MEM43; MEM43 antigen; membrane attack complex (MAC) inhibition factor; membrane attack complex inhibition factor; Membrane inhibitor of reactive lysis; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20; protectin; surface anitgen recognized by monoclonal antibody 16.3A5; T cell-activating protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.