Learn More
CD59 Mouse anti-Human, Brilliant Violet 605, Clone: p282 (H19), BD Optibuild™
Shop All BD Biosciences ProductsDescription
The p282 (HI9) monoclonal antibody specifically binds to CD59, a 19 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein, expressed on hematopoietic and non-hematopoietic cells. Because of its interaction with complement activated products, CD59 has been termed membrane-attack-complex-inhibitory factor (MACIF), homologus restriction factor (HRF20), membrane inhibitor of reactive lysis (MIRL) and Protectin. It inhibits the cytolytic activity of the complement system by binding to C8 and C9, thereby blocking the assembly of the membrane attack complex. CD59 also participates in spontaneous T-cell/erythrocyte adhesion, interacts with CD2, and plays a role in T-cell activation.
Specifications
Specifications
| Antigen | CD59 |
| Classification | Monoclonal |
| Clone | p282 (H19) |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 605 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | HRF-20, MAC-inhibitory protein, MAC-IP, MACIF, MEM43, MIRL, Protectin, 1F5 |
| Host Species | Mouse |
| Immunogen | Human Erythrocytes |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.