Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD59 Overexpression Lysate
SDP

Catalog No. NBP210121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP210121 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP210121 Supplier Novus Biologicals™ Supplier No. NBP210121
Add to Cart
Add to Cart
The CD59 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_97675. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens CD59, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Synonym 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.