Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD59 Rabbit anti-Human, Clone: 29, Novus Biologicals™

Rabbit Monoclonal Antibody
$366.00 - $572.50
Specifications
| Antigen | CD59 |
|---|---|
| Clone | 29 |
| Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
CD59 Monoclonal specifically detects CD59 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
| CD59 | |
| Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein | |
| Recombinant Human CD59 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 29 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 966 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title