Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD59 Recombinant Rabbit Monoclonal Antibody (029), PE
GREENER_CHOICE

Catalog No. PIMA546756
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546756 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546756 Supplier Invitrogen™ Supplier No. MA546756
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD59 (Met1-Glu101) and conjugated with PE under optimum conditions, the unreacted PE was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

CD59 (Protectin) is a small (18-20 kDa) GPI-anchored ubiquitously expressed inhibitor of the membrane attack complex (MAC). CD59 is the key regulator that preserves the autologous cells from terminal effector mechanism of the complement cascade. CD59 also associates with C5b-8 complex and counteracts appropriate formation of cytolytic pore within the plasma membrane. Further, CD59 is a low-affinity ligand of human CD2, causes T cell costimulation, and is involved in lymphocyte signal transduction. CD59 is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. Mutations in this gene cause CD59 deficiency, a disease involving hemolytic anemia, thrombosis, and cerebral infarction. Multiple alternatively spliced transcript variants of CD59 have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD59
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 029
Concentration 100 μg/mL
Conjugate PE
Formulation PBS with 0.5% BSA and 0.09% sodium azide
Gene CD59
Gene Accession No. P13987
Gene Alias 16.3A5; 1F5; 1F5 antigen; 20 kDa homologous restriction factor; AA987121; Cb59b molecule; CD59; CD59 antigen; CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344); CD59 glycoprotein; CD59 molecule; CD59 molecule (CD59 blood group); CD59 molecule, complement regulatory protein; Cd59a; CD59a antigen; CD59A glycoprotein; Cd59b; CD59b antigen; CD59B glycoprotein; CD59b moleucle, complement regulatory protein; complement inhibitor CD59; complement regulatory protein; EJ16; EJ30; EL32; G344; HRF20; HRF-20; human leukocyte antigen MIC11; Ly-6-like protein; lymphocytic antigen CD59/MEM43; MACIF; MAC-inhibitory protein; MACIP; MAC-IP; MEM43; MEM43 antigen; membrane attack complex (MAC) inhibition factor; membrane attack complex inhibition factor; Membrane inhibitor of reactive lysis; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20; protectin; surface anitgen recognized by monoclonal antibody 16.3A5; T cell-activating protein
Gene Symbols CD59
Host Species Rabbit
Immunogen Recombinant protein ecoding Human CD59 (Met1-Glu101).
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 966
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.