Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5L Antibody (CD5L/2991), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279841PCP
Description
CD5L Monoclonal specifically detects CD5L in Human samples. It is validated for Flow Cytometry.Specifications
CD5L | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
CD5L | |
Recombinant fragment of human CD5L protein (around aa 269-366) (exact sequence is proprietary) (Uniprot: O43866) | |
0.1 mL | |
Apoptosis | |
Human | |
IgG2b κ |
Flow Cytometry | |
CD5L/2991 | |
Flow Cytometry | |
AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction