Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5L/Api6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984763
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4763 Supplier Bioss Supplier No. BS2487R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apoptosis inhibitor of macrophages (AIM) is a member of the scavenger receptor cysteine-rich domain superfamily (SRCR-SF) initially identified as an inducible cell surface ligand of CD5. It was shown that AIM functions in the thymus as the inducer of resistance to apoptosis within CD4+/CD8+ thymocytes and as the supporter of the viability of these cells before thymic selection. AIM was also shown to support macrophage survival and enhance their phagocytic function. More recent experiments using recombinant AIM significantly inhibited apoptosis of NKT and T cells obtained from C. parvum-stimulated livers in vitro, suggesting that AIM functions to induce resistance to apoptosis in these cells and supports host defense against inflammation during infection.

Specifications

Antigen CD5L/Api6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cd5l
Gene Accession No. O43866
Gene Alias 1/6; AAC-11; AI047839; Aim; AIM/Spalpha; API6; apoptosis inhibitor 6; Apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; Apoptosis inhibitory 6; CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; CD5 molecule-like; CD5L; CT2; CT-2; hAIM; IgM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; Spalpha; SP-alpha; UNQ203/PRO229
Gene Symbols Cd5l
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CD5L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 922
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.