Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD6 Antibody (SPV-L14), DyLight 755 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311571IR
Description
CD6 Monoclonal antibody specifically detects CD6 in Human, Mouse samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD6 | |
Monoclonal | |
DyLight 755 | |
CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
CD8+ cytotoxic T-cell clone | |
0.1 mL | |
Cytokine Research, Immunology, Signal Transduction | |
923 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
SPV-L14 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction