Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD6 Monoclonal Antibody (6B9)
GREENER_CHOICE

Catalog No. PIMA517047
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517047 Supplier Invitrogen™ Supplier No. MA517047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17047 targets CD6 in IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17047 immunogen is purified recombinant fragment of human CD6 (amino acids: 472-668) expressed in E. Coli. MA5-17047 detects CD6 which has a predicted molecular weight of approximately 71.8kDa.

CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CD6
Gene Accession No. P30203
Gene Alias Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120
Gene Symbols CD6
Host Species Mouse
Immunogen Purified recombinant fragment of human CD6 (amino acids: 472-668) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 923
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.