Learn More
Invitrogen™ CD6 Monoclonal Antibody (MEM-98), PE

Description
This antibody reacts with an extracellular epitope of CD6, a 100-130 kDa single chain transmembrane glycoprotein expressed on T and B lymphocytes subsets, thymocytes, and acute lymphocytic leukemia cells.
Specifications
Specifications
| Antigen | CD6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-98 |
| Concentration | 0.03 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | CD6 |
| Gene Accession No. | P30203 |
| Gene Alias | Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.