Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6 Mouse, anti-Mouse, R718, Clone: J90-462, BD OptiBuild™

Catalog No. BDB751956 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751956 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751956 Supplier BD Biosciences Supplier No. 751956
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The J90-462 monoclonal antibody specifically recognizes CD6. CD6 is a 105-130 kDa type I transmembrane glycoprotein that belongs to the scavenger receptor family. It is expressed on most thymocytes and peripheral T cells. CD6 is highly expressed on CD3-high thymocytes and expressed at lower levels on CD4+ CD8+ double positive and CD4+CD8- or CD4-CD8+ single positive thymocytes. CD6 is also reportedly expressed at low levels by some neurons. CD6 serves as a receptor for CD166, also known as Activated Leukocyte Cell Adhesion Molecule (ALCAM). CD6 plays a role in adhesion and migration during organogenesis. The CD6 intracellular domain is constitutively phosphorylated which suggests that CD6 may play a signaling role in T cell activation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD6
Applications Flow Cytometry
Classification Monoclonal
Clone J90-462
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.