Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117117
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117117 Supplier Invitrogen™ Supplier No. PA5117117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Jurkat, U-937, HT-29, Raji Immunogen sequence: LRIKGKYALP VMVNHQHLPT TIPAGSNSYQ PVPITIPKEV FMLPIQVQAP PPEDSDSGSD SDYEHYDFSA QPPVALTTFY NSQRHRVTDE EVQQSRFQMP PLEEGLEELH ASHIPTANPG HCITDPPSLG PQYHPRSNSE SSTSSGEDYC NSPKSKLPPW NPQVFSSERS SFLEQPPNLE LAGTQPAFSA GPPADDSSST SSGEWYQNFQ PPPQPPSEEQ FGCPGSPSPQ PDSTDNDDYD DISAA.

CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD6
Gene Accession No. P30203
Gene Alias Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120
Gene Symbols CD6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 424-668 of human CD6 (NP_0067163).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 923
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.