Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD6 Rabbit anti-Mouse, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29069150UL
Description
CD6 Monoclonal specifically detects CD6 in Mouse samples. It is validated for Sandwich ELISA Detection.Specifications
CD6 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
2 | |
Sandwich ELISA Detection 1:1000-1:10000 | |
CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
Recombinant Mouse CD6 Protein | |
50 μg | |
Cytokine Research, Immunology, Signal Transduction | |
923 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction