Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Hamster anti-Mouse, BB700, Clone: 2C9.G2 (also known as HMβ3-1), BD Optibuild™

Catalog No. BDB742110 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742110 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742110 Supplier BD Biosciences Supplier No. 742110
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The 2C9.G2 monoclonal antibody specifically binds to the integrin β3 chain (CD61), which associates with the integrin αv chain (CD51) to form the vitronectin receptor, as well as the αIIb chain (CD41) to form the gpIIb/IIIa complex. Both receptors mediate adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, and von Willebrand factor. Leukocyte-endothelial adhesion is also mediated by the binding of αvβ3 integrin or vitronectin receptor to CD31 (PECAM-1). In addition, interaction of the αvβ3 integrin with its ligands regulates the L-type Ca2+ channel in vascular smooth muscle cells, possibly mediating vasodilatory responses to injury. Soluble and insoluble 2C9.G2 mAb mimics the effect of the natural ligands in smooth muscle cells from rat cremaster arterioles. Furthermore, osteopontin, also named Eta-1, is a cytokine that binds to αvβ3. CD61 is expressed on platelets, activated T lymphocytes, polymorphonuclear granulocytes, and blastocysts. Cross-reactivity of mAb 2C9.G2 to rat mast cells and platelets has been observed by flow cytometric analysis. mAb 2C9.G2 has been demonstrated to block binding of rat and mouse cells to fibronectin

The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5.  Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61
Classification Monoclonal
Clone 2C9.G2 (also known as HMβ3-1)
Concentration 0.2mg/mL
Conjugate Brilliant Blue 700 (BB700)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Itgb3, Integrin beta-3, INGRB3, Platelet glycoprotein IIIa, GP3A, GPIIIa
Host Species Armenian Hamster
Immunogen Mouse T-cell Hybridoma 2B4 Vitronectin Receptor
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.