Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD61 (Integrin beta 3) Monoclonal Antibody (2C9.G3), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$201.00 - $379.00
Specifications
Antigen | CD61 (Integrin beta 3) |
---|---|
Clone | 2C9.G3 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M053T03Y04A
![]() |
Invitrogen™
M053T03Y04A |
100 μg |
Each for $379.00
|
|
|||||
M053T02Y04A
![]() |
Invitrogen™
M053T02Y04A |
25 μg |
Each for $201.00
|
|
|||||
Description
CD61 (Integrin beta 3) Monoclonal antibody specifically detects CD61 (Integrin beta 3) in Mouse, Rat samples. It is validated for Flow CytometrySpecifications
CD61 (Integrin beta 3) | |
0.1 mg/mL | |
Monoclonal | |
Armenian Hamster | |
Mouse, Rat | |
O54890 | |
16416, 29302 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
2C9.G3 | |
Flow Cytometry | |
NovaFluor Yellow 660 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
IgG | |
Affinity chromatography | |
ITGB3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title