Learn More
CD61 Monoclonal Antibody (43.PL), PE, Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0614100TESTS


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD61 Monoclonal antibody specifically detects CD61 in Human samples. It is validated for Flow CytometrySpecifications
CD61 | |
Monoclonal | |
PE | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P05106 | |
Mouse | |
Protein A/G | |
RUO | |
3690 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry | |
43.PL | |
100UG HU FOXP3 PCH101 AFP647 MAB 100 UG | |
ITGB3 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
CD61 was derived from the hybridization of mouse X63.Ag8.653 myeloma cells with spleen cells of BALB/c mice immunized with purified platelet membrane glycoproteins. | |
100Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.