Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Mouse anti-Human, Brilliant Violet 421, Clone: VI-PL2 (also known as VIPL2), BD Optibuild™

Catalog No. BDB744381 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744381 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744381 Supplier BD Biosciences Supplier No. 744381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The VI-PL2 monoclonal antibody specifically binds to CD61. CD61 is a 105 kDa transmembrane glycoprotein that is also known as integrin 3 and platelet glycoprotein IIIa (GPIIIa or GP3A). It is expressed on platelets, megakaryocytes, osteoclasts and endothelia. Integrin 3 associates with gpIIa (CD41) to form the CD41/CD61 complex which mediates platelet adhesion and aggregation. CD61 also associates with CD51 to form the CD51/CD61 complex (vitronectin receptor). CD61 appears to bind to fibrinogen, fibronectin, vWF, vitronectin, and thrombospondin to mediate cell adhesion.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61
Classification Monoclonal
Clone VI-PL2 (also known as VIPL2)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin β3, Integrin beta-3, GP3A, GPIIIa, ITGB3, ITB3
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.