Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Mouse anti-Rat, FITC, Clone: F11, BD

Catalog No. BDB561909 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561909 50 μg
BDB554952 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561909 Supplier BD Biosciences Supplier No. 561909
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The F11 antibody reacts with the integrin β3 chain (CD61), which associates with the integrin αv chain (CD51), to form the vitronectin receptor found on endothelial cells, myeloid cells, and osteoclasts, and with the αIIb chain (CD41) on platelets and megakaryocytes. Both receptors mediate adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, and von Willebrand factor. F11 mAb has been reported to block the in vitro attachment of osteoclasts to several ligands. Weak reactivity with human osteoclasts, megakaryocytes, and platelets has also been reported.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61
Applications Flow Cytometry
Classification Monoclonal
Clone F11
Concentration 0.5mg/mL
Conjugate FITC
Description Integrin β3 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat (Sprague-Dawley) bone cell suspensions
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.