Learn More
Invitrogen™ CD62E/CD62P Monoclonal Antibody (1.2B6), Biotin

Description
Clone 1.2B6 recognizes a common epitope shared by human CD62P and CD62E.
Specifications
Specifications
| Antigen | CD62E/CD62P |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1.2B6 |
| Concentration | 0.1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | SELE |
| Gene Accession No. | P16109, P16581, P98110 |
| Gene Alias | CD62; CD62 antigen-like family member E; CD62 antigen-like family member P; CD62E; CD62P; CD62P; CD62 homolog; ELAM; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule 1; ESEL; E-selectin; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; GRMP; LECAM2; LECAM-2; LECAM3; LECAM-3; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; P-selectin; P-selectin precursor; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); SELP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.