Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62E Mouse anti-Human, Brilliant Violet 786, Clone: 68-5H11, BD Optibuild™

Catalog No. BDB742707 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742707 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742707 Supplier BD Biosciences Supplier No. 742707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 68-5H11 monoclonal antibody specifically binds to CD62E. This adhesion molecule is a 97-115 kDa type I transmembrane glycoprotein that is encoded by the SELE /gene. CD62E is also known as E-selectin, Endothelial-leukocyte adhesion molecule-1 (ELAM-1), and Leukocyte endothelial cell adhesion molecule 2 (LECAM-2). CD62E is minimally expressed by unstimulated endothelium. Activated endothelial cells upregulate surface CD62E expression in response to various activators including inflammatory cytokines such as Interleukin-1 and Tumor Necrosis Factor. CD62E plays a role in leucocyte extravasation by promoting leucocyte rolling on activated endothelial cells during inflammation. CD62E may also play a role in the metastasis of certain tumor cells. The adhesion between endothelial CD62E molecules and a carbohydrate ligand on neutrophils is inhibited by the mAb 68-5H11.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62E
Classification Monoclonal
Clone 68-5H11
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SELE, E-selectin, Selectin E, ELAM, ELAM1, ESEL, LECAM2
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.