Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD62L (L-Selectin) Monoclonal Antibody (DREG-56 (DREG56)), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD62L (L-Selectin) |
---|---|
Clone | DREG-56 (DREG56) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H009T03B09A
![]() |
Invitrogen™
H009T03B09A |
100 Tests |
Each for $430.00
|
|
|||||
H009T02B09A
![]() |
Invitrogen™
H009T02B09A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Human samples. It is validated for Flow CytometrySpecifications
CD62L (L-Selectin) | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
P14151 | |
6402 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
DREG-56 (DREG56) | |
Flow Cytometry | |
NovaFluor Blue 690 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
IgG1 κ | |
Affinity chromatography | |
SELL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title