Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD62L (L-Selectin) Monoclonal Antibody (MEL-14), NovaFluor™ Yellow 590, eBioscience™

Catalog No. M006T02Y02A
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M006T02Y02A 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M006T02Y02A Supplier Invitrogen™ Supplier No. M006T02Y02A
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MEL-14 monoclonal antibody reacts with mouse CD62L, a 76 kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCam-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues. This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer. Applications Tested The MEL-14 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.2 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Master mixes Master mixes of NFs should be made at 2-8 °C and may be made up to 4 hours ahead of time. We do not recommend storing master mixes containing NovaFluor conjugates overnight or longer. Whole Blood compatibility When utilizing whole blood (as opposed to density-gradient-purified PBMC), we recommend lysing red blood cells in bulk prior t...

The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62L (L-Selectin)
Applications Flow Cytometry
Classification Monoclonal
Clone MEL-14
Concentration 0.1 mg/mL
Conjugate NovaFluor Yellow 590
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene SELL
Gene Accession No. P18337
Gene Alias A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1
Gene Symbols SELL
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20343
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.