Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L (L-Selectin), Mouse Anti-Human, RB613, Clone: DREG-56, BD OptiBuild™

Catalog No. BDB759482 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg;
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB759482 50 μg;
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB759482 Supplier BD Biosciences Supplier No. 759482
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Human (Tested in Development) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62L (L-Selectin)
Applications Flow Cytometry
Classification Monoclonal
Clone DREG-56
Concentration 0.2 mg/mL
Conjugate RB613
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1
Host Species Mouse
Immunogen Supernatant from PMA-activated Human Peripheral Blood Leukocytes
Quantity 50 μg;
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6402
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.