Learn More
Invitrogen™ CD62L Monoclonal Antibody (1H3)

Description
CD62L Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD62L |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1H3 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | SELL |
| Gene Accession No. | P14151 |
| Gene Alias | A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 |
| Gene Symbols | SELL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.